WHEATON® E-Z Vial® with Snap Ring, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
225179-03 225179 225179-01 225179-02
66014-582CS 806.95 CAD
66014-582 89235-542 89235-544 89235-546
WHEATON® E-Z Vial® with Snap Ring, DWK Life Sciences
Vials
Clear or amber Type I borosilicate glass.
  • Snap ring vials eliminate the need for crimping and decapping tools
  • Vials accept snap cap or 11 mm aluminum seal
  • 40% larger opening improves sample accessibility while reducing auto sampler needle damage