Spectra/Por® 4, Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Supplier: Spectrum Laboratories
132700 132703 132709 132706 132697 S632706
25219-220EA 631.94 CAD
25219-220 25219-223 25219-230 74400-004 25224-375 28170-160
Spectra/Por® 4, Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Tubing Dialysis Tubing
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes.
  • Hydrophilic, symmetric porosity
Tubing is packaged dry for handling and storage convenience. The tubing is available in 15 m/roll and 30 m/roll to provide better economy for applications requiring larger amounts of tubing.
Dry without glycerin and no soaking is required.