Microlink® Screw Caps, WHEATON
Supplier: DWK Life Sciences (KIMBLE)
240235-01 240106 240114 240200 240112 240250-01 240256-01
14227-558CS 312.06 CAD
14227-558 16299-210 16299-216 16199-935 16299-214 80086-296 89508-346
Microlink® Screw Caps, WHEATON
Closures Bottle Closures
These screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
  • Liner is ultrasonically welded to the cap to eliminate the possibility of glue contamination and bond failure common with autoclaving
  • Available in convenient lab pack quantities as well as larger case packs
  • Low-compression-set silicone ideal for the storage of samples for prolonged period of time over wide temperature ranges
  • PTFE or polypropylene on the exposed face of the liner imparts an inert,chemical-resistant surface
  • Open top caps provide access with a syringe
  • Autoclavable