Simultaneous Test Sets, Drager Safety
Supplier: Draeger
8101735 8101770 8101736
CA13453-124PK 636.2 CAD
CA13453-124 CA13453-128 13453-126
Simultaneous Test Sets, Drager Safety
Gas Detection Systems
Risks and potential dangers at a glance: The SimultanTest Sets allow the measurement of five different gases at the same time.
In the SimultanTest Set, five Dräger-Tubes are arranged next to each other in a stable rubber sleeve. The air to be monitored is simultaneously drawn in through the tubes by a Dräger-Tube pump. Quick determinations are made about the presence of dangerous substances at TLV levels.
The device delivers fast and reliable results for different combinations of organic and inorganic gases.