Reagent Bottles, with Screw Cap, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
220163 220223 220283
15507-164CS 143.74 CAD
15507-164 15507-168 15507-170
Reagent Bottles, with Screw Cap, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Made of borosilicate glass.
  • Features no-drip pour lip
  • Pre-attached polypropylene screw cap with PTFE liner
  • Large writing patch
  • Autoclavable