WHEATON® Crimpers and Decappers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
224322 224323 224307 224316 224357 224321
14217-400CS 1303.62 CAD
14217-400 14217-402 26676-406 26676-413 26676-426 10816-282
WHEATON® Crimpers and Decappers, DWK Life Sciences
Crimpers, Cappers and Decappers
These crimpers are used for flip cap seals and for aluminum seals sizes 16.5, 28.5, and 30 mm.
  • 1 year warranty
  • Not autoclavable