Potassium 98%, chunks in mineral oil

Supplier: Thermo Scientific Chemicals
Danger

L13267-14 L13267-22
AAL13267-14EA 280.72 CAD
AAL13267-14 AAL13267-22
Potassium 98%, chunks in mineral oil
Potassium

Potassium, chunks, in mineral oil, 98%


Purity: 98% CAS No.: 7440-09-7 Molecular Formula: K Formula Weight: 39.10 Physical Form: Solid Density: 0.860 Appearance: Silver/grey Odor: Odorless Melting Point: 63-64° Boiling Point: 765-770° Merck Reference: 14,7604 UN No.: 2257 MDL No.: MFCD00133776

Formula: K
Boiling Pt: 765…770 °C
Melting Pt: 63…64 °C
Density: 0.860
Storage Temperature: Ambient
MDL Number: MFCD00133776
CAS Number: 7440-09-7
EINECS: 231-119-8
UN: 2257
ADR: 4.3,I
Merck Index: 14,7604

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR