Propanephosphonic acid cyclic anhydride ≥50% (w/w) in acetonitrile

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Propylphosphonic anhydride, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide

H66967-18 H66967-30
AAH66967-18EA 109.18 CAD
AAH66967-18 AAH66967-30
Propanephosphonic acid cyclic anhydride ≥50% (w/w) in acetonitrile
Propanephosphonic acid cyclic anhydride

1-Propylphosphonic acid cyclic anhydride, 50+% w/w soln. in acetonitrile


CAS No.: 68957-94-8 Molecular Formula: C9H21O6P3 Formula Weight: 318.19 Physical Form: Liquid Appearance: Colorless to brown Beilstein Reference: 5079255 UN No.: 2924 MDL No.: MFCD00006583

Formula: C₉H₂₁O₆P₃
MW: 318.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006583
CAS Number: 68957-94-8
EINECS: 422-210-5
UN: 2924
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR