2-Bromo-3'-chloropropiophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64999-03
H64999-06
AAH64999-03EA
59.25
CAD
AAH64999-03
AAH64999-06
2-Bromo-3'-chloropropiophenone 98%
2-Bromo-3'-chloropropiophenone
2-Bromo-3'-chloropropiophenone, 98%
Purity: 98%
CAS No.: 34911-51-8
Molecular Formula: C9H8BrClO
Formula Weight: 247.52
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.532
Refractive Index: 1.5770
Appearance: Colorless to pale yellow
Beilstein Reference: 2360404
MDL No.: MFCD03701150
Formula:
C₉H₈BrClO Density: 1.532 Storage Temperature: Ambient |
MDL Number:
MFCD03701150 CAS Number: 34911-51-8 EINECS: 252-282-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...