2-Bromo-3-nitro-6-picoline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64982-06
H64982-14
AAH64982-03EA
68.08
CAD
AAH64982-03
AAH64982-06
AAH64982-14
2-Bromo-3-nitro-6-picoline 98%
2-Bromo-3-nitro-6-picoline
2-Bromo-6-methyl-3-nitropyridine, 98%
Purity: 98%
CAS No.: 374633-31-5
Molecular Formula: C6H5BrN2O2
Formula Weight: 217.02
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 138200
MDL No.: MFCD03095212
Formula:
C₆H₅BrN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03095212 CAS Number: 374633-31-5 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...