4-Bromoimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

H64972-06 H64972-14
AAH64972-06EA 92.38 CAD
AAH64972-06 AAH64972-14
4-Bromoimidazole 98%
4-Bromoimidazole

4-Bromoimidazole, 98%


Purity: 98% CAS No.: 2302-25-2 Molecular Formula: C3H3BrN2 Formula Weight: 146.97 Physical Form: Solid Melting Point: 131-135° Beilstein Reference: 1615 UN No.: 2811 MDL No.: MFCD00047021

Formula: C₃H₃BrN₂
Melting Pt: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00047021
CAS Number: 2302-25-2
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR