Copper(I)-2-thiophenecarboxylate
Supplier: Thermo Scientific Chemicals
|
Danger
|
H64950-03
H64950-06
AAH64950-03EA
137.14
CAD
AAH64950-03
AAH64950-06
Copper(I)-2-thiophenecarboxylate
Copper(I)-2-thiophenecarboxylate
Copper(I) thiophene-2-carboxylate
CAS No.: 68986-76-5
Molecular Formula: C5H3CuO2S
Formula Weight: 190.68
Physical Form: Powder
Sensitivity: Air and Light Sensitive
Appearance: Red to brown
MDL No.: MFCD02183524
Formula:
C₅H₃CuO₂S Storage Temperature: Ambient |
MDL Number:
MFCD02183524 CAS Number: 68986-76-5 UN: 3077 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...