2-Chloro-5-pyrimidineboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloropyrimidine-5-boronic acid

H64949-03 H64949-06
AAH64949-03EA 108.7 CAD
AAH64949-03 AAH64949-06 AAH64949-MD
2-Chloro-5-pyrimidineboronic acid 96%
2-Chloro-5-pyrimidineboronic acid

2-Chloropyrimidine-5-boronic acid, 96%


Purity: 96% CAS No.: 1003845-06-4 Molecular Formula: C4H4BClN2O2 Formula Weight: 158.35 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD08063113

Formula: C₄H₄BClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08063113
CAS Number: 1003845-06-4

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR