3-Cyanophenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester
H64948-03
H64948-06
AAH64948-03EA
58.14
CAD
AAH64948-03
AAH64948-06
3-Cyanophenylboronic acid pinacol ester 97%
3-Cyanophenylboronic acid pinacol ester
3-Cyanobenzeneboronic acid pinacol ester, 97%
Purity: 97%
CAS No.: 214360-46-0
Molecular Formula: C13H16BNO2
Formula Weight: 229.09
Physical Form: Solid
Appearance: White
Melting Point: 78-82° dec.
MDL No.: MFCD03789262
Formula:
C₁₃H₁₆BNO₂ MW: 229.09 g/mol Melting Pt: 78…82 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03789262 CAS Number: 214360-46-0 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...