5-(Trifluoromethyl)picolinic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(Trifluoromethyl)pyridine-2-carboxylic acid

H64937-03 H64937-06 H64937-14
AAH64937-03EA 69.84 CAD
AAH64937-03 AAH64937-06 AAH64937-14
5-(Trifluoromethyl)picolinic acid 95%
5-(Trifluoromethyl)picolinic acid

5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%


Purity: 95% CAS No.: 80194-69-0 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Physical Form: Powder Appearance: Pale brown Melting Point: 133-137° MDL No.: MFCD04113632

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD04113632
CAS Number: 80194-69-0

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR