dibenzo[b,d]furan-2-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

H64905-03 H64905-06
AAH64905-03EA 164.32 CAD
AAH64905-03 AAH64905-06 AAH64905-MD
dibenzo[b,d]furan-2-carbaldehyde 98%
dibenzo[b,d]furan-2-carbaldehyde

Dimethyl pyrimidine-2,5-dicarboxylate, 97%


Purity: 98% CAS No.: 5397-82-0 Molecular Formula: C13H8O2 Formula Weight: 196.20 Physical Form: Powder Sensitivity: Air Sensitive Appearance: Off-white Melting Point: 74° Beilstein Reference: 141044 MDL No.: MFCD00136272

Formula: C₁₃H₈O₂
Melting Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00136272
CAS Number: 5397-82-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR