3-Bromo-5-chlorophenol 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64844-06
H64844-14
AAH64844-03EA
38.93
CAD
AAH64844-03
AAH64844-06
AAH64844-14
3-Bromo-5-chlorophenol 97+%
3-Bromo-5-chlorophenol
3-Bromo-5-chlorophenol, 97+%
Purity: 97+%
CAS No.: 56962-04-0
Molecular Formula: C6H4BrClO
Formula Weight: 207.45
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 66-70°
Beilstein Reference: 3234780
MDL No.: MFCD07780676
Formula:
C₆H₄BrClO MW: 207.45 g/mol Melting Pt: 66…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780676 CAS Number: 56962-04-0 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...