4-(Trifluoromethyl)piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Trifluoromethylpiperidine

H64840-03 H64840-06 H64840-MD
AAH64840-03EA 237 CAD
AAH64840-03 AAH64840-06 AAH64840-MD
4-(Trifluoromethyl)piperidine 97%
4-(Trifluoromethyl)piperidine

4-(Trifluoromethyl)piperidine, 97%


Purity: 97% CAS No.: 657-36-3 Molecular Formula: C6H10F3N Formula Weight: 153.15 Physical Form: Liquid Appearance: Colorless MDL No.: MFCD00102068

Formula: C₆H₁₀F₃N
MW: 153.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00102068
CAS Number: 657-36-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR