4-Nitroanthranilic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

H64831-14 H64831-22
AAH64831-14EA 72.05 CAD
AAH64831-14 AAH64831-22 AAH64831-36
4-Nitroanthranilic acid 97+%
4-Nitroanthranilic acid

2-Amino-4-nitrobenzoic acid, 97+%


Purity: 97+% CAS No.: 619-17-0 Molecular Formula: C7H6N2O4 Formula Weight: 182.13 Physical Form: Solid Melting Point: 257° lit. Beilstein Reference: 785071 MDL No.: MFCD00007262

Formula: C₇H₆N₂O₄
Melting Pt: 257 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00007262
CAS Number: 619-17-0
EINECS: 210-583-5

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR