8-Bromo-1-octene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64830-03
H64830-06
AAH64830-03EA
70.78
CAD
AAH64830-03
AAH64830-06
8-Bromo-1-octene 97%
8-Bromo-1-octene
8-Bromo-1-octene, 97%
Purity: 97%
CAS No.: 2695-48-9
Molecular Formula: C8H15Br
Formula Weight: 191.11
Flash Point: 78°(172°F)
Physical Form: Liquid
Density: 1.139
Refractive Index: 1.4670
Appearance: Pale yelllow
Boiling Point: 92°/24mm
Beilstein Reference: 2234481
MDL No.: MFCD00000275
Formula:
C₈H₁₅Br Boiling Pt: 92 °C/24 mm Density: 1.139 Flash Pt: 78 °C(172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000275 CAS Number: 2695-48-9 EINECS: 220-268-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...