1-Bromo-5-fluoropentane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64806-03
H64806-06
H64806-MD
AAH64806-03EA
139.2
CAD
AAH64806-03
AAH64806-06
AAH64806-MD
1-Bromo-5-fluoropentane 99%
1-Bromo-5-fluoropentane
1-Bromo-5-fluoropentane, 99%
Purity: 99%
CAS No.: 407-97-6
Molecular Formula: C5H10BrF
Formula Weight: 169.04
Physical Form: Liquid
Density: 1.360
Refractive Index: 1.4406
Appearance: Colorless
Boiling Point: 162°
Beilstein Reference: 1697258
MDL No.: MFCD01709395
Formula:
C₅H₁₀BrF Boiling Pt: 162 °C Density: 1.360 Storage Temperature: Ambient |
MDL Number:
MFCD01709395 CAS Number: 407-97-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...