Methyl-2-chloronicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-chloronicotinate, Methyl 2-chloropyridine-3-carboxylate

H64798-06 H64798-14 H64798-22
AAH64798-06EA 43.61 CAD
AAH64798-06 AAH64798-14 AAH64798-22
Methyl-2-chloronicotinate 98%
Methyl 2-chloronicotinate

Methyl 2-chloropyridine-3-carboxylate, 98%


Purity: 98% CAS No.: 40134-18-7 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.314 Refractive Index: 1.5370 Appearance: Colorless Boiling Point: 150°/1mm Beilstein Reference: 122129 MDL No.: MFCD01318629

Formula: C₇H₆ClNO₂
MW: 171.58
Boiling Pt: 150 °C/1 mm
Density: 1.314
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01318629
CAS Number: 40134-18-7
EINECS: 254-805-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR