9-Bromo-1-nonene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64792-03
H64792-06
AAH64792-03EA
55.77
CAD
AAH64792-03
AAH64792-06
9-Bromo-1-nonene 97%
9-Bromo-1-nonene
9-Bromo-1-nonene, 97%
Purity: 97%
CAS No.: 89359-54-6
Molecular Formula: C9H17Br
Formula Weight: 205.14
Physical Form: Liquid
Refractive Index: 1.4675
Appearance: Colorless to pale yellow
Boiling Point: 100-101°/15mm
Beilstein Reference: 1746338
MDL No.: MFCD09037826
Formula:
C₉H₁₇Br Boiling Pt: 100…101 °C/15 mm Storage Temperature: Ambient |
MDL Number:
MFCD09037826 CAS Number: 89359-54-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...