3-Bromo-5-nitrobenzaldehyde 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64787-03
H64787-06
AAH64787-03EA
123.08
CAD
AAH64787-03
AAH64787-06
AAH64787-MD
3-Bromo-5-nitrobenzaldehyde 97+%
3-Bromo-5-nitrobenzaldehyde
3-Bromo-5-nitrobenzaldehyde, 97+%
Purity: 97+%
CAS No.: 355134-13-3
Molecular Formula: C7H4BrNO3
Formula Weight: 230.02
Physical Form: Powder
Appearance: Pale brown
Melting Point: 101-106°
Beilstein Reference: 3253229
MDL No.: MFCD00100097
Formula:
C₇H₄BrNO₃ Melting Pt: 101…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100097 CAS Number: 355134-13-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...