2-Chloro-5-methylpyrimidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64773-06
H64773-14
AAH64773-03EA
42.08
CAD
AAH64773-03
AAH64773-06
AAH64773-14
2-Chloro-5-methylpyrimidine 97%
2-Chloro-5-methylpyrimidine
2-Chloro-5-methylpyrimidine, 97%
Purity: 97%
CAS No.: 22536-61-4
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Physical Form: Solid
Melting Point: 89-92°
Beilstein Reference: 108732
MDL No.: MFCD09260903
Formula:
C5H5ClN2 Melting Pt: 89…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD09260903 CAS Number: 22536-61-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...