4-Bromo-1-methyl-1H-pyrazole 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H64768-14
H64768-22
AAH64768-14EA
101.59
CAD
AAH64768-14
AAH64768-22
4-Bromo-1-methyl-1H-pyrazole 98+%
4-Bromo-1-methyl-1H-pyrazole
4-Bromo-1-methyl-1H-pyrazole, 98+%
Purity: 98+%
CAS No.: 15803-02-8
Molecular Formula: C4H5BrN2
Formula Weight: 161.00
Flash Point: 94°(201°F)
Physical Form: Liquid
Density: 1.558
Refractive Index: 1.5310
Appearance: Colorless to pale yellow
Boiling Point: 185-188°
Beilstein Reference: 108120
MDL No.: MFCD02179565
Formula:
C₄H₅BrN₂ Boiling Pt: 185…188 °C Density: 1.558 Flash Pt: 94 °C(201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02179565 CAS Number: 15803-02-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...