Methyl-5-bromo-6-chloronicotinate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64754-06
H64754-14
AAH64754-03EA
40.04
CAD
AAH64754-03
AAH64754-06
AAH64754-14
Methyl-5-bromo-6-chloronicotinate 98%
Methyl 5-bromo-6-chloronicotinate
Methyl 5-bromo-6-chloronicotinate, 98%
Purity: 98%
CAS No.: 78686-77-8
Molecular Formula: C7H5BrClNO2
Formula Weight: 250.48
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 76-79°
Beilstein Reference: 4403950
MDL No.: MFCD03844855
Formula:
C₇H₅BrClNO₂ Melting Pt: 76…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD03844855 CAS Number: 78686-77-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...