2-Bromo-9,9-dimethylfluorene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64732-06
H64732-14
H64732-22
AAH64732-06EA
68.73
CAD
AAH64732-06
AAH64732-14
AAH64732-22
2-Bromo-9,9-dimethylfluorene 98%
2-Bromo-9,9-dimethylfluorene
2-Bromo-9,9-dimethylfluorene, 98%
Purity: 98%
CAS No.: 28320-31-2
Molecular Formula: C15H13Br
Formula Weight: 273.17
Physical Form: Solid
Melting Point: 57-62°
Beilstein Reference: 2524870
MDL No.: MFCD09261272
Formula:
C₁₅H₁₃Br MW: 273.17 g/mol Melting Pt: 57…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD09261272 CAS Number: 28320-31-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...