4-(Trifluoromethyl)-1H-imidazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Trifluoromethyl)imidazole
H64711-03
H64711-06
AAH64711-03EA
68.26
CAD
AAH64711-03
AAH64711-06
AAH64711-MD
4-(Trifluoromethyl)-1H-imidazole 98%
4-(Trifluoromethyl)-1H-imidazole
4-(Trifluoromethyl)imidazole, 98%
Purity: 98%
CAS No.: 33468-69-8
Molecular Formula: C4H3F3N2
Formula Weight: 136.08
Physical Form: Solid
Melting Point: 148-150°
MDL No.: MFCD08458860
Formula:
C4H3F3N2 MW: 136.08 g/mol Melting Pt: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458860 CAS Number: 33468-69-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...