4-Bromo-1H-pyrazole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64673-14
H64673-36
AAH64673-14EA
82.32
CAD
AAH64673-14
AAH64673-36
AAH64673-A1
4-Bromo-1H-pyrazole 98+%
4-Bromo-1H-pyrazole
4-Bromo-1H-pyrazole, 98+%
Purity: 98+%
CAS No.: 2075-45-8
Molecular Formula: C3H3BrN2
Formula Weight: 146.97
Physical Form: Solid
Melting Point: 93-96° lit.
Boiling Point: 250-260° lit.
Beilstein Reference: 106510
MDL No.: MFCD00075602
Formula:
C₃H₃BrN₂ Boiling Pt: 250…260° lit. Melting Pt: 93…96 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00075602 CAS Number: 2075-45-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...