2-Bromodibenzothiophene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromodibenzo[b,d]thiophene

H64645-03 H64645-06 H64645-14
AAH64645-03EA 60.67 CAD
AAH64645-03 AAH64645-06 AAH64645-14
2-Bromodibenzothiophene 98%
2-Bromodibenzothiophene

2-Bromodibenzothiophene, 98%


Purity: 98% CAS No.: 22439-61-8 Molecular Formula: C12H7BrS Formula Weight: 263.15 Melting Point: 123-129° lit Beilstein Reference: 142203 MDL No.: MFCD00089285

Formula: C₁₂H₇BrS
MW: 263.15 g/mol
Melting Pt: 123…129 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00089285
CAS Number: 22439-61-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR