2-Bromo-4,5-difluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64621-03
H64621-06
AAH64621-03EA
124.19
CAD
AAH64621-03
AAH64621-06
AAH64621-MD
2-Bromo-4,5-difluorobenzaldehyde 98%
2-Bromo-4,5-difluorobenzaldehyde
2-Bromo-4,5-difluorobenzaldehyde, 98%
Purity: 98%
CAS No.: 476620-54-9
Molecular Formula: C7H3BrF2O
Formula Weight: 221.00
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 20365202
MDL No.: MFCD16659662
Formula:
C₇H₃BrF₂O Storage Temperature: Ambient |
MDL Number:
MFCD16659662 CAS Number: 476620-54-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...