2-Bromo-6-chlorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64605-03
H64605-06
AAH64605-03EA
169.06
CAD
AAH64605-03
AAH64605-06
AAH64605-MD
2-Bromo-6-chlorobenzaldehyde 98%
2-Bromo-6-chlorobenzaldehyde
2-Bromo-6-chlorobenzaldehyde, 98%
Purity: 98%
CAS No.: 64622-16-8
Molecular Formula: C7H4BrClO
Formula Weight: 219.46
Physical Form: Solid
Melting Point: 81-86°
Beilstein Reference: 2613087
MDL No.: MFCD00266792
Formula:
C₇H₄BrClO Melting Pt: 81…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00266792 CAS Number: 64622-16-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...