Tetrahydropyran-4-methanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H64568-03
H64568-06
AAH64568-03EA
95.91
CAD
AAH64568-03
AAH64568-06
AAH64568-MD
Tetrahydropyran-4-methanol 98%
Tetrahydropyran-4-methanol
4-(Hydroxymethyl)tetrahydropyran, 98%
Purity: 98%
CAS No.: 14774-37-9
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 61°(142°F)
Physical Form: Liquid
Refractive Index: 1.4600
Boiling Point: 111-115°/15mm
Beilstein Reference: 103069
MDL No.: MFCD00457804
Formula:
C₆H₁₂O₂ Boiling Pt: 111…115 °C/15 mm Flash Pt: 61 °C(142 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00457804 CAS Number: 14774-37-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...