2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64551-06
H64551-14
AAH64551-06EA
95.27
CAD
AAH64551-06
AAH64551-14
AAH64551-22
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine 97+%
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine
2,3-Dihydro-7-azaindole, 97+%
Purity: 97+%
CAS No.: 10592-27-5
Molecular Formula: C7H8N2
Formula Weight: 120.15
Physical Form: Powder
Appearance: White
Melting Point: 78-83°
Beilstein Reference: 116366
MDL No.: MFCD06659751
Formula:
C7H8N2 Melting Pt: 78…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659751 CAS Number: 10592-27-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...