Isopropyl benzenesulfonate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64523-03
H64523-06
H64523-14
AAH64523-03EA
73.63
CAD
AAH64523-03
AAH64523-06
AAH64523-14
Isopropyl benzenesulfonate 95%
Isopropyl benzenesulfonate
Benzenesulfonic acid isopropyl ester, 95%
Purity: 95%
CAS No.: 6214-18-2
Molecular Formula: C9H12O3S
Formula Weight: 200.25
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Refractive Index: 1.5020
Appearance: Colorless to pale yellow
Beilstein Reference: 1106314
MDL No.: MFCD04038771
Formula:
C₉H₁₂O₃S Storage Temperature: Ambient |
MDL Number:
MFCD04038771 CAS Number: 6214-18-2 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...