N-(4-Oxocyclohexyl)acetamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64511-06
H64511-14
H64511-22
AAH64511-06EA
45.35
CAD
AAH64511-06
AAH64511-14
AAH64511-22
N-(4-Oxocyclohexyl)acetamide 98%
N-(4-Oxocyclohexyl)acetamide
N-(4-Oxocyclohexyl)acetamide, 98%
Purity: 98%
CAS No.: 27514-08-5
Molecular Formula: C8H13NO2
Formula Weight: 155.20
Physical Form: Solid
Appearance: Off-white
Melting Point: 136-140°
Beilstein Reference: 2803083
MDL No.: MFCD03703462
Formula:
C8H13NO2 Melting Pt: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD03703462 CAS Number: 27514-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...