2,6-Dimethylbenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64503-06
H64503-14
AAH64503-06EA
68.41
CAD
AAH64503-06
AAH64503-14
AAH64503-22
2,6-Dimethylbenzaldehyde 97%
2,6-Dimethylbenzaldehyde
2,6-Dimethylbenzaldehyde, 97%
Purity: 97%
CAS No.: 1123-56-4
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 96°(205°F)
Physical Form: Solid
Refractive Index: 1.5480
Sensitivity: Air Sensitive
Appearance: White to yellow
Melting Point: 27-30° lit.
Boiling Point: 230° lit.
Beilstein Reference: 907019
MDL No.: MFCD00128003
Formula:
C₉H₁₀O Boiling Pt: 230° (lit.) Melting Pt: 27…30 °C (lit.) Flash Pt: 96 °C(205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00128003 CAS Number: 1123-56-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...