3-Methoxyphenethylamine 97+%

Supplier: Thermo Scientific Chemicals
Danger

H64494-06 H64494-14 H64494-22
AAH64494-06EA 65.38 CAD
AAH64494-06 AAH64494-14 AAH64494-22
3-Methoxyphenethylamine 97+%
3-Methoxyphenethylamine

2-(3-Methoxyphenyl)ethylamine, 97+%


Purity: 97+% CAS No.: 2039-67-0 Molecular Formula: C9H13NO Formula Weight: 151.21 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.038 Refractive Index: 1.5380 Appearance: Yellow Boiling Point: 118-118°/6mm Beilstein Reference: 775202 UN No.: 2735 MDL No.: MFCD00008187

Formula: C₉H₁₃NO
Boiling Pt: 118…118 °C/6 mm
Density: 1.038
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008187
CAS Number: 2039-67-0
EINECS: 218-017-9
UN: 2735
ADR: "8",III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR