1,5-Naphthyridine 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H64484-03
H64484-06
H64484-MD
AAH64484-03EA
233.84
CAD
AAH64484-03
AAH64484-06
AAH64484-MD
1,5-Naphthyridine 97+%
1,5-Naphthyridine
1,5-Naphthyridine, 97+%
Purity: 97+%
CAS No.: 254-79-5
Molecular Formula: C8H6N2
Formula Weight: 130.15
Physical Form: Solid
Density: 1.25
Appearance: Off-white
Melting Point: 69-75°
Beilstein Reference: 109350
MDL No.: MFCD00039727
Formula:
C₈H₆N₂ Melting Pt: 69…75 °C Density: 1.25 Storage Temperature: Ambient |
MDL Number:
MFCD00039727 CAS Number: 254-79-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...