3-Amino-2-pyridone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol, 3-Amino-2-pyridinone
H64480-03
H64480-06
AAH64480-03EA
135.72
CAD
AAH64480-03
AAH64480-06
AAH64480-MD
3-Amino-2-pyridone 98%
3-Amino-2-pyridone
3-Amino-2-hydroxypyridine, 98%
Purity: 98%
CAS No.: 33630-99-8
Molecular Formula: C5H6N2O
Formula Weight: 110.12
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline solid
Appearance: Brown
Beilstein Reference: 109849
MDL No.: MFCD03411556
Formula:
C₅H₆N₂O MW: 110.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03411556 CAS Number: 33630-99-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...