Benzyl-3-oxopiperidine-1-carboxylate 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Benzyloxycarbonyl-3-piperidone, Benzyl 3-oxopiperidine-1-carboxylate
H64471-03
H64471-06
H64471-14
AAH64471-03EA
88.95
CAD
AAH64471-03
AAH64471-06
AAH64471-14
Benzyl-3-oxopiperidine-1-carboxylate 97+%
Benzyl-3-oxopiperidine-1-carboxylate
1-Benzyloxycarbonyl-3-piperidone, 97+%
Purity: 97+%
CAS No.: 61995-20-8
Molecular Formula: C13H15NO3
Formula Weight: 233.27
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Beilstein Reference: 1464624
MDL No.: MFCD03001723
Formula:
C13H15NO3 MW: 233.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001723 CAS Number: 61995-20-8 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...