2-Isopropoxy-5-pyridineboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64461-06
H64461-MD
AAH64461-03EA
169.76
CAD
AAH64461-03
AAH64461-06
AAH64461-MD
2-Isopropoxy-5-pyridineboronic acid 97%
2-Isopropoxy-5-pyridineboronic acid
2-Isopropoxypyridine-5-boronic acid, 97%
Purity: 97%
CAS No.: 870521-30-5
Molecular Formula: C8H12BNO3
Formula Weight: 181.00
Physical Form: Solid
MDL No.: MFCD07368870
Formula:
C₈H₁₂BNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD07368870 CAS Number: 870521-30-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...