5-Bromo-2-chlorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

H64438-03 H64438-06
AAH64438-03EA 70.31 CAD
AAH64438-03 AAH64438-06
5-Bromo-2-chlorobenzaldehyde 98%
5-Bromo-2-chlorobenzaldehyde

5-Bromo-2-chlorobenzaldehyde, 98%


Purity: 98% CAS No.: 189628-37-3 Molecular Formula: C7H4BrClO Formula Weight: 219.46 Physical Form: Solid Appearance: Off-white to pale yellow Beilstein Reference: 9757388 MDL No.: MFCD08445659

Formula: C₇H₄BrClO
Storage Temperature: Ambient
MDL Number: MFCD08445659
CAS Number: 189628-37-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR