2,3-Dimethylbenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
H64425-06
H64425-14
H64425-22
AAH64425-06EA
54.51
CAD
AAH64425-06
AAH64425-14
AAH64425-22
2,3-Dimethylbenzaldehyde 97%
2,3-Dimethylbenzaldehyde
2,3-Dimethylbenzaldehyde, 97%
Purity: 97%
CAS No.: 5779-93-1
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 101°(214°F)
Physical Form: Liquid
Density: 1.029
Refractive Index: 1.5530
Appearance: Colorless to pale yellow
Boiling Point: 86-88°/10mm
Beilstein Reference: 2238677
MDL No.: MFCD00798004
Formula:
C₉H₁₀O Boiling Pt: 86…88 °C/10 mm Density: 1.029 Flash Pt: 101 °C(214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00798004 CAS Number: 5779-93-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...