2-Bromo-3-fluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64400-06
H64400-14
H64400-22
AAH64400-06EA
93.85
CAD
AAH64400-06
AAH64400-14
AAH64400-22
2-Bromo-3-fluorobenzaldehyde 98%
2-Bromo-3-fluorobenzaldehyde
2-Bromo-3-fluorobenzaldehyde, 98%
Purity: 98%
CAS No.: 891180-59-9
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Beilstein Reference: 10529128
MDL No.: MFCD07782044
Formula:
C₇H₄BrFO Storage Temperature: Ambient |
MDL Number:
MFCD07782044 CAS Number: 891180-59-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...