Benzo[b]furan-3-boronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64389-03
H64389-06
H64389-MD
AAH64389-03EA
413.96
CAD
AAH64389-03
AAH64389-06
AAH64389-MD
Benzo[b]furan-3-boronic acid 98%
Benzo[b]furan-3-boronic acid
Benzo[b]furan-3-boronic acid, 98%
Purity: 98%
CAS No.: 317830-83-4
Molecular Formula: C8H7BO3
Formula Weight: 161.95
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 9479377
MDL No.: MFCD06801687
Formula:
C₈H₇BO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD06801687 CAS Number: 317830-83-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...