L-Lysine ethyl ester diisocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H64384-06
H64384-14
AAH64384-06EA
206.98
CAD
AAH64384-06
AAH64384-14
L-Lysine ethyl ester diisocyanate 97%
L-Lysine ethyl ester diisocyanate
L-Lysine ethyl ester diisocyanate, 97%
Purity: 97%
CAS No.: 45172-15-4
Molecular Formula: C10H14N2O4
Formula Weight: 226.23
Storage Temperature: 2°C to 8°C
Physical Form: Viscous liquid
Refractive Index: 1.4572
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Optical Rotation: -35° (c=1 in dioxane)
MDL No.: MFCD08276480
Formula:
C₁₀H₁₄N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD08276480 CAS Number: 45172-15-4 UN: 2206 ADR: "6.1",III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...