2-Bromo-5-chloropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64367-14
H64367-22
AAH64367-14EA
85.64
CAD
AAH64367-14
AAH64367-22
2-Bromo-5-chloropyridine 98%
2-Bromo-5-chloropyridine
2-Bromo-5-chloropyridine, 98%
Purity: 98%
CAS No.: 40473-01-6
Molecular Formula: C5H3BrClN
Formula Weight: 192.44
Physical Form: Solid
Melting Point: 65-69° lit.
Boiling Point: 128°/16mm
Beilstein Reference: 109098
MDL No.: MFCD00234006
Formula:
C₅H₃BrClN Boiling Pt: 128 °C/16 mm Melting Pt: 65…69 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00234006 CAS Number: 40473-01-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...