4-Amino-5-bromopyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64335-03
H64335-06
AAH64335-03EA
191.18
CAD
AAH64335-03
AAH64335-06
AAH64335-MD
4-Amino-5-bromopyrimidine 98%
4-Amino-5-bromopyrimidine
4-Amino-5-bromopyrimidine, 98%
Purity: 98%
CAS No.: 1439-10-7
Molecular Formula: C4H4BrN3
Formula Weight: 174.00
Physical Form: Powder
Appearance: Pale brown
Melting Point: 207-211°
Beilstein Reference: 112648
MDL No.: MFCD00233962
Formula:
C₄H₄BrN₃ Melting Pt: 207…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233962 CAS Number: 1439-10-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...